Assay Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025517.3 | NP_079793.2 | 7 - 8 | 928 | 61 | |
| GenBank mRNA | AK002543.1 | - | 7 - 8 | 838 | 61 | |
| AK009245.1 | - | 7 - 8 | 864 | 61 | ||
| AK151108.1 | - | 7 - 8 | 816 | 61 | ||
| AK151400.1 | - | 7 - 8 | 816 | 61 | ||
| AK152129.1 | - | 7 - 8 | 929 | 61 | ||
| AK159961.1 | - | 7 - 8 | 818 | 61 | ||
| AK167830.1 | - | 7 - 8 | 805 | 61 | ||
| AK190960.1 | - | 1 - 2 | 84 | 61 | ||
| AK193515.1 | - | 1 - 2 | 65 | 61 | ||
| AK207601.1 | - | 1 - 2 | 65 | 61 | ||
| AK209245.1 | - | 1 - 2 | 65 | 61 | ||
| AK219075.1 | - | 1 - 2 | 65 | 61 | ||
| BC016519.1 | - | 7 - 8 | 854 | 3495638 | 61 | |
| BC090403.1 | - | 7 - 8 | 899 | 5293337 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rtca
Genomic Map