Assay Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025616.3 | NP_079892.1 | 9 - 10 | 864 | 69 | |
| GenBank mRNA | AK008340.1 | - | 9 - 10 | 865 | 69 | |
| AK008382.1 | - | 9 - 10 | 861 | 69 | ||
| AK010832.1 | - | 9 - 10 | 830 | 69 | ||
| AK088074.1 | - | 9 - 10 | 855 | 69 | ||
| AK167473.1 | - | 9 - 10 | 867 | 69 | ||
| AY551342.1 | - | 9 - 10 | 850 | 69 | ||
| BC010303.1 | - | 9 - 10 | 839 | 3597676 | 69 | |
| BC025844.1 | - | 3 - 4 | 345 | 69 | ||
| BC037056.1 | - | 9 - 10 | 866 | 69 | ||
More Information
Set Membership: |
|

Timm50
Genomic Map