Assay Details
| Gene Symbol: Ogfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025671.2 | NP_079947.1 | 4 - 5 | 398 | 74 | |
| GenBank mRNA | AK004916.1 | - | 2 - 3 | 866 | 74 | |
| AK004943.1 | - | 4 - 5 | 399 | 74 | ||
| AK017497.1 | - | 4 - 5 | 399 | 74 | ||
| AK018534.1 | - | 4 - 5 | 399 | 74 | ||
| AK168946.1 | - | 4 - 5 | 407 | 74 | ||
| BC022762.1 | - | 3 - 4 | 246 | 74 | ||
| BC069872.1 | - | 4 - 5 | 401 | 4219293 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC022762.1 transcripts.
Set Membership: |
|

Ogfod2
Genomic Map