Assay Details
| Gene Symbol: Ogfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025671.2 | NP_079947.1 | 6 - 7 | 787 | 86 | |
| GenBank mRNA | AK004916.1 | - | 3 - 4 | 1450 | 86 | |
| AK004943.1 | - | 6 - 7 | 788 | 86 | ||
| AK017497.1 | - | 6 - 7 | 787 | 85 | ||
| AK018534.1 | - | 6 - 7 | 788 | 86 | ||
| AK168946.1 | - | 6 - 7 | 705 | 86 | ||
| BC022762.1 | - | 5 - 6 | 635 | 86 | ||
| BC069872.1 | - | 6 - 7 | 790 | 4219293 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oxidation-reduction process
sulfonate dioxygenase activity
molecular_function
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
L-ascorbic acid binding
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
oxidation-reduction process
sulfonate dioxygenase activity
molecular_function
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
L-ascorbic acid binding
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059331 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977353 | BC022762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977353 | AK168946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977353 | AK004943.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977353 | AK017497.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977353 | AK018534.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900292 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29317 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119120 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977353 | BC022762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977353 | AK168946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977353 | AK004943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977353 | AK017497.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977353 | AK018534.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24059356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119120 | - | - |
| Whole Mouse Genome G4122A | A_51_P228974 | BC022762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P228974 | AK168946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P228974 | AK004943.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P228974 | AK017497.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P228974 | AK018534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P128951 | BC022762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P128951 | BC069872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P128951 | AK168946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P128951 | AK004916.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P128951 | AK004943.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121063 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115591 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119120 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04356371 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04691965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04219048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04219041 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04691958 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04356370 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04356379 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04219043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04356373 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04356380 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04691957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24059335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24059383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04356371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04356373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04356379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04356370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04219053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04359315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04356380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24059341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525713 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429238_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525711 | - | - |
| Mouse Genome 430 2.0 Array | 1429238_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429238_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66627_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66627_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500012870.mm.1 | TC0500001436.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012870.mm.1 | NM_025671.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012870.mm.1 | NM_025671.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012873.mm.1 | TC0500001436.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012873.mm.1 | NM_025671.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012873.mm.1 | NM_025671.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012871.mm.1 | TC0500001436.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012871.mm.1 | NM_025671.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012871.mm.1 | NM_025671.2 | Refseq |
| Murine Genome U74v2 Set | 160922_at | - | - |
| Murine Genome U74v2 Set | 160922_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429238_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429238_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6860575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6860575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228590 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228590 | - | - |
| mouse-6 v1.1 (Illumina) | scl27225.5.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27225.5.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228590 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2732896 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732896 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228590 | - | - |

Ogfod2
Genomic Map