Assay Details
| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025819.4 | NP_080095.3 | 5 - 6 | 455 | 145 | |
| GenBank mRNA | AK004803.2 | - | 5 - 6 | 402 | 145 | |
| AK013686.1 | - | 5 - 6 | 400 | 145 | ||
| BC028449.1 | - | 5 - 6 | 383 | 145 | ||
| BC055064.1 | - | 5 - 6 | 456 | 6528686 | 145 | |
| BC060204.1 | - | 5 - 6 | 395 | 6855442 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Swt1
Genomic Map