Assay Details
| Gene Symbol: Naa16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025832.2 | NP_080108.1 | 13 - 14 | 1589 | 138 | |
| GenBank mRNA | AK005056.1 | - | 13 - 14 | 1599 | 138 | |
| AK007755.1 | - | 13 - 14 | 1625 | 138 | ||
| AK028432.1 | - | 13 - 14 | 1700 | 138 | ||
| AK152498.1 | - | 13 - 14 | 1599 | 138 | ||
| BC052445.1 | - | 13 - 14 | 1589 | 6412745 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa16
Genomic Map