Assay Details
| Gene Symbol: Naa16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025832.2 | NP_080108.1 | 17 - 18 | 2210 | 113 | |
| GenBank mRNA | AK005056.1 | - | 17 - 18 | 2220 | 113 | |
| AK028432.1 | - | 17 - 18 | 2321 | 113 | ||
| AK152498.1 | - | 17 - 18 | 2220 | 113 | ||
| BC048727.1 | - | 5 - 6 | 751 | 113 | ||
| BC052445.1 | - | 17 - 18 | 2210 | 6412745 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa16
Genomic Map