Assay Details
| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164082.1 | NP_001157554.1 | 5 - 6 | 766 | 83 | |
| NM_025945.3 | NP_080221.3 | 6 - 7 | 798 | 83 | ||
| GenBank mRNA | AK013165.1 | - | 5 - 6 | 683 | 83 | |
| AK156587.1 | - | 6 - 7 | 750 | 83 | ||
| BC016102.1 | - | 6 - 7 | 774 | 4911475 | 83 | |
More Information
Set Membership: |
|

Polr3d
Genomic Map