Assay Details
| Gene Symbol: Ccdc47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026009.2 | NP_080285.2 | 1 - 2 | 215 | 76 | |
| GenBank mRNA | AB040489.1 | - | 1 - 2 | 192 | 76 | |
| AK011882.1 | - | 1 - 2 | 152 | 76 | ||
| AK078812.1 | - | 1 - 2 | 194 | 76 | ||
| AK132273.1 | - | 1 - 2 | 173 | 76 | ||
| BC120502.1 | - | 1 - 2 | 103 | 40129425 | 76 | |
| BC137681.1 | - | 1 - 2 | 103 | 8860700 | 76 | |
| CJ157793.1 | - | 1 - 2 | 215 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB040489.1,AK011882.1,AK078812.1,AK132273.1,BC120502.1,BC137681.1,NM_026009.2 transcripts.
Set Membership: |
|

Ccdc47
Genomic Map