Assay Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025582.2 | NP_001020753.1 | 3 - 4 | 394 | 64 | |
| NM_001286986.1 | NP_001273915.1 | 3 - 4 | 391 | 64 | ||
| NM_001286987.1 | NP_001273916.1 | 2 - 3 | 269 | 64 | ||
| NM_026013.3 | NP_080289.1 | 3 - 4 | 394 | 64 | ||
| GenBank mRNA | AI875295.1 | - | 3 - 4 | 394 | 64 | |
| AK008532.1 | - | 3 - 4 | 318 | 64 | ||
| AK009940.1 | - | 3 - 4 | 331 | 64 | ||
| AK012044.1 | - | 3 - 4 | 311 | 64 | ||
| AK015888.1 | - | 3 - 4 | 357 | 64 | ||
| AK052824.1 | - | 2 - 3 | 203 | 64 | ||
| AK172455.1 | - | 3 - 4 | 361 | 64 | ||
| BC024458.1 | - | 3 - 4 | 320 | 4975302 | 64 | |
| BI738450.1 | - | 3 - 4 | 362 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22402369 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22402370 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22402368 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980746 | AK172455.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980746 | BC024458.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980746 | AK012044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980746 | AK015888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980746 | AK009940.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86088 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81152 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125526 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151468 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980746 | AK172455.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980746 | BC024458.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980746 | AK012044.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980746 | AK015888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980746 | AK009940.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22402389 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00720304 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151468 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00720312 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125526 | - | - |
| Whole Mouse Genome G4122A | A_51_P380801 | AK172455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380801 | BC024458.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380801 | AK012044.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380801 | AK052824.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P380801 | NM_001025582.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120520 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00720304 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125526 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151468 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02545951 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02545940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02545944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22402370 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22402369 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22402368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22402415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02673354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02744481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22402382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495166 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445545_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424782_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453731_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495163 | - | - |
| Mouse Genome 430 2.0 Array | 1453731_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1453731_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424782_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424782_at | - | - |
| Mouse Genome 430 2.0 Array | 1445545_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445545_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67171_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67171_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300008989.mm.1 | NM_001025582.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008989.mm.1 | NM_001286986.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008989.mm.1 | NM_001286987.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008989.mm.1 | NM_026013.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008989.mm.1 | TC0300001104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300008987.mm.1 | NM_001025582.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008987.mm.1 | NM_001286986.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008987.mm.1 | NM_001286987.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008987.mm.1 | NM_026013.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008987.mm.1 | TC0300001104.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424782_at | - | - |
| Mouse Expression Set 430 | 1453731_PM_a_at | - | - |
| Mouse Expression Set 430 | 1453731_a_at | - | - |
| Mouse Expression Set 430 | 1424782_PM_at | - | - |
| Mouse Expression Set 430 | 1445545_PM_at | - | - |
| Mouse Expression Set 430 | 1445545_at | - | - |
| Murine Genome U74v2 Set | 161540_r_at | - | - |
| Murine Genome U74v2 Set | 161540_PM_r_at | - | - |
| Murine Genome U74v2 Set | 96688_at | - | - |
| Murine Genome U74v2 Set | 96688_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453731_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424782_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424782_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453731_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424782_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424782_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453731_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453731_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445545_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445545_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100430368 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224589 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224589 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2489135 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067171.2_285-S | - | - |
| mouse-6 v1.1 (Illumina) | scl22747.11_296-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110324 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100430368 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067171.2_285-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100430368 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3051608 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1224589 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3127932 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3051608 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224589 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3127932 | - | - |

Dram2
Genomic Map