Assay Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286986.1 | NP_001273915.1 | 6 - 7 | 772 | 108 | |
| NM_001286987.1 | NP_001273916.1 | 5 - 6 | 650 | 108 | ||
| NM_026013.3 | NP_080289.1 | 6 - 7 | 775 | 108 | ||
| GenBank mRNA | AK009940.1 | - | 6 - 7 | 712 | 108 | |
| AK012044.1 | - | 6 - 7 | 692 | 108 | ||
| AK015888.1 | - | 6 - 7 | 738 | 108 | ||
| AK052824.1 | - | 5 - 6 | 584 | 108 | ||
| AK172455.1 | - | 6 - 7 | 742 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dram2
Genomic Map