Assay Details
| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026192.3 | NP_080468.1 | 5 - 6 | 771 | 91 | |
| GenBank mRNA | AK007393.1 | - | 5 - 6 | 662 | 91 | |
| AK018734.2 | - | 5 - 6 | 657 | 91 | ||
| AK035598.1 | - | 5 - 6 | 683 | 91 | ||
| AK087513.1 | - | 5 - 6 | 725 | 91 | ||
| AK220400.1 | - | 5 - 6 | 684 | 91 | ||
| AY131329.1 | - | 4 - 5 | 637 | 91 | ||
| BC054783.1 | - | 5 - 6 | 665 | 6815943 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Calcoco1
Genomic Map