Assay Details
| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026192.3 | NP_080468.1 | 13 - 14 | 1943 | 81 | |
| GenBank mRNA | AK007393.1 | - | 13 - 14 | 1834 | 81 | |
| AK018734.2 | - | 13 - 14 | 1830 | 81 | ||
| AK035598.1 | - | 13 - 14 | 1855 | 81 | ||
| AK087513.1 | - | 13 - 14 | 1897 | 81 | ||
| AK220400.1 | - | 13 - 14 | 1856 | 81 | ||
| AY131329.1 | - | 12 - 13 | 1809 | 81 | ||
| BC054783.1 | - | 13 - 14 | 1837 | 6815943 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Calcoco1
Genomic Map