Assay Details
| Gene Symbol: Mettl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026197.3 | NP_080473.1 | 3 - 4 | 453 | 81 | |
| GenBank mRNA | AK011780.1 | - | 3 - 4 | 454 | 81 | |
| AK012739.1 | - | 3 - 4 | 454 | 81 | ||
| AK013306.1 | - | 3 - 4 | 465 | 81 | ||
| AK029045.1 | - | 3 - 4 | 437 | 81 | ||
| AK145022.1 | - | 3 - 4 | 461 | 81 | ||
| AK145662.1 | - | 3 - 4 | 453 | 81 | ||
| AK168798.1 | - | 3 - 4 | 403 | 81 | ||
| BC003467.1 | - | 3 - 4 | 424 | 81 | ||
| BC022902.1 | - | 3 - 4 | 465 | 81 | ||
| BC037452.1 | - | 3 - 4 | 402 | 81 | ||
| BC049897.1 | - | 3 - 4 | 382 | 3979938 | 81 | |
| BC054724.1 | - | 3 - 4 | 389 | 5695738 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl16
Genomic Map