Assay Details
| Gene Symbol: Gpr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026229.4 | NP_080505.1 | 12 - 13 | 1165 | 98 | |
| GenBank mRNA | AK016787.1 | - | 12 - 13 | 1165 | 98 | |
| AK034931.1 | - | 831 | 98 | |||
| BC010729.1 | - | 12 - 13 | 1158 | 3708143 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr89
Genomic Map