Assay Details
| Gene Symbol: Spc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026282.5 | NP_080558.1 | 1 - 2 | 168 | 105 | |
| GenBank mRNA | AK011728.1 | - | 1 - 2 | 186 | 105 | |
| AK031680.1 | - | 1 - 2 | 165 | 105 | ||
| AK075950.1 | - | 1 - 2 | 169 | 105 | ||
| AK153185.1 | - | 1 - 2 | 186 | 105 | ||
| AK167877.1 | - | 1 - 2 | 188 | 105 | ||
| BC056965.1 | - | 1 - 2 | 173 | 105 | ||
| BC086683.1 | - | 1 - 2 | 171 | 30676712 | 105 | |
| BC116238.1 | - | 1 - 2 | 169 | 40091783 | 105 | |
| BC116239.1 | - | 1 - 2 | 168 | 40091784 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348780 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348781 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348778 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348779 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056473 | BC098472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056473 | BC116238.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056473 | BC116239.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056473 | AK031680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056473 | AK075950.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443512 | BC119062.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443512 | BC120746.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443512 | NM_011195.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443512 | U16958.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263794 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111789 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105558 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111520 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178403 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178296 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139532 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056473 | BC098472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056473 | BC116238.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056473 | BC116239.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056473 | AK031680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056473 | AK075950.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26348787 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06719493 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111520 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103110 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01882645 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06719117 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06719151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178296 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01882581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01882412 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06719127 | - | - |
| Whole Mouse Genome G4122A | A_51_P214209 | AK031680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214209 | AK075950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214209 | NM_026282.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214209 | AK011728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214209 | BC098472.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113987 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178296 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103110 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06719127 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111520 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01882581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07738460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07510746 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07889027 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07982429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07982427 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07982433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348779 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348781 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348780 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348778 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26348782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26348767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07990010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07982429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07879678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07706473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07982433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07988699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07829904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07854107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08062077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08061521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26348774 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591562 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591559 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591560 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591558 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431087_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591556 | - | - |
| Mouse Genome 430 2.0 Array | 1431087_at | - | - |
| Mouse Genome 430 2.0 Array | 1431087_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67629_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67629_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900015800.mm.1 | NM_026282.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015800.mm.1 | NM_026282.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015800.mm.1 | TC0900001883.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900015801.mm.1 | NM_026282.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015801.mm.1 | NM_026282.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015801.mm.1 | TC0900001883.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431087_at | - | - |
| Mouse Expression Set 430 | 1431087_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431087_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431087_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 619055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4280451 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1940563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2750890 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2615709 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2615709 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2750890 | - | - |
| mouse-6 v1.1 (Illumina) | scl067629.2_23-S | - | - |
| mouse-6 v1.1 (Illumina) | scl36134.1.46_72-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940563 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36134.1.46_72-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067629.2_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940563 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280451 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2615709 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2948272 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2948272 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2615709 | - | - |

Spc24
Genomic Map