Assay Details
| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026360.3 | NP_080636.2 | 6 - 7 | 657 | 86 | |
| GenBank mRNA | AK010310.1 | - | 6 - 7 | 650 | 86 | |
| AK076982.1 | - | 6 - 7 | 642 | 86 | ||
| AK169595.1 | - | 4 - 5 | 280 | 86 | ||
| BC060168.1 | - | 6 - 7 | 637 | 86 | ||
| BC095944.1 | - | 6 - 7 | 634 | 4935970 | 86 | |
| BC132242.1 | - | 6 - 7 | 649 | 40130519 | 86 | |
| BC132244.1 | - | 6 - 7 | 649 | 40130521 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx47
Genomic Map