Assay Details
| Gene Symbol: S100a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026416.2 | NP_080692.1 | 2 - 3 | 275 | 68 | |
| GenBank mRNA | AK009762.1 | - | 2 - 3 | 226 | 68 | |
| AK160178.1 | - | 2 - 3 | 226 | 68 | ||
| AK160512.1 | - | 2 - 3 | 218 | 68 | ||
| AK167006.1 | - | 2 - 3 | 218 | 68 | ||
| AY221962.1 | - | 1 - 2 | 166 | 68 | ||
| BC020031.1 | - | 2 - 3 | 275 | 3980274 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114627 | AK160178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114627 | AK160512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114627 | AK167006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114627 | BC020031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114627 | BC032079.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148912 | AK160178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148912 | AK160512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148912 | AK167006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148912 | BC020031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148912 | BC032079.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264958 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169006 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114627 | AK160178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114627 | AK160512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114627 | AK167006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114627 | BC020031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114627 | BC032079.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148912 | AK160178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148912 | AK160512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148912 | AK167006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148912 | BC020031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148912 | BC032079.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22325798 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169006 | - | - |
| Whole Mouse Genome G4122A | A_51_P134533 | AK160512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P134533 | AK167006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P134533 | AY221962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P134533 | BC020031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P134533 | NM_026416.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114627 | AK160178.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114627 | AK160512.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114627 | AK167006.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114627 | BC020031.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114627 | BC032079.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02990428 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02990420 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02990416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02442810 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02990430 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02990422 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02990424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02442813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22325785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03156492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02817508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02442820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02990419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02990420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03092220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02442819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02990416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02366440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02990428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02990422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02442810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22325772 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493802 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493801 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447676_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425560_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493798 | - | - |
| Mouse Genome 430 2.0 Array | 1447676_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447676_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1425560_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425560_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67860_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67860_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006276.mm.1 | NM_026416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006276.mm.1 | NM_026416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006276.mm.1 | TC0300000802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006270.mm.1 | NM_026416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006270.mm.1 | NM_026416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006270.mm.1 | TC0300000802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006275.mm.1 | NM_026416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006275.mm.1 | NM_026416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006275.mm.1 | TC0300000802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006271.mm.1 | NM_026416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006271.mm.1 | NM_026416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006271.mm.1 | TC0300000802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006273.mm.1 | NM_026416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006273.mm.1 | NM_026416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006273.mm.1 | TC0300000802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006274.mm.1 | NM_026416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006274.mm.1 | NM_026416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006274.mm.1 | TC0300000802.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425560_a_at | - | - |
| Mouse Expression Set 430 | 1425560_PM_a_at | - | - |
| Mouse Expression Set 430 | 1447676_x_at | - | - |
| Mouse Expression Set 430 | 1447676_PM_x_at | - | - |
| Murine Genome U74v2 Set | 95747_PM_at | - | - |
| Murine Genome U74v2 Set | 95747_at | - | - |
| Murine Genome U74v2 Set | 167148_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167148_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425560_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425560_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425560_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425560_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447676_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447676_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2510546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002510546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220521 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220521 | - | - |
| mouse-6 v1.1 (Illumina) | scl22945.5.27_48-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2510546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22945.5.27_48-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220521 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220521 | - | - |

S100a16
Genomic Map