Assay Details
| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163388.1 | NP_001156860.1 | 1 - 2 | 155 | 84 | |
| NM_026511.2 | NP_080787.1 | 1 - 2 | 155 | 84 | ||
| GenBank mRNA | AK002873.1 | - | 1 - 2 | 157 | 84 | |
| AK005403.1 | - | 1 - 2 | 143 | 84 | ||
| AK010493.1 | - | 1 - 2 | 145 | 84 | ||
| AK012648.1 | - | 1 - 2 | 153 | 84 | ||
| AK029408.1 | - | 1 - 2 | 131 | 84 | ||
| AK148446.1 | - | 1 - 2 | 95 | 84 | ||
| AK160910.1 | - | 1 - 2 | 139 | 84 | ||
| BC071262.1 | - | 1 - 2 | 155 | 6509633 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of smooth muscle cell apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
molecular_function
apoptotic process
positive regulation of smooth muscle cell apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745655 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745656 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745654 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060445 | BC071262.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060445 | BY392793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060445 | AK010493.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060445 | AK012648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060445 | AK029408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060444 | BC071262.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060444 | BY392793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060444 | AK012648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060444 | AK029408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060444 | NM_026511.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795721 | AK038689.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795721 | AK049160.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563785 | AK042350.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795510 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67590 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163004 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130865 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060444 | BC071262.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060444 | BY392793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060444 | AK012648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060444 | AK029408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060444 | NM_026511.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060445 | BC071262.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060445 | BY392793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060445 | AK010493.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060445 | AK012648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060445 | AK029408.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02608110 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130865 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163004 | - | - |
| Whole Mouse Genome G4122A | A_52_P451334 | AK047131.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P112066 | BC071262.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P112066 | AK002873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P112066 | AK005403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P112066 | AK010493.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P112066 | AK012648.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122144 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130865 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09884574 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10116075 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745656 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745654 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10108752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10121952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28745659 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398725 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454309_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440183_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429248_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398721 | - | - |
| Mouse Genome 430 2.0 Array | 1454309_at | - | - |
| Mouse Genome 430 2.0 Array | 1440183_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1440183_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454309_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429248_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429248_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68020_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68020_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200008856.mm.1 | NM_001163388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008856.mm.1 | NM_001163388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008856.mm.1 | NM_026511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008856.mm.1 | NM_026511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008856.mm.1 | TC1200001244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200008853.mm.1 | NM_001163388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008853.mm.1 | NM_001163388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008853.mm.1 | NM_026511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008853.mm.1 | NM_026511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008853.mm.1 | TC1200001244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200008866.mm.1 | NM_026511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008866.mm.1 | NM_026511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008866.mm.1 | TC1200001244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200008857.mm.1 | NM_001163388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008857.mm.1 | NM_001163388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008857.mm.1 | NM_026511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008857.mm.1 | NM_026511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200008857.mm.1 | TC1200001244.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454309_at | - | - |
| Mouse Expression Set 430 | 1454309_PM_at | - | - |
| Mouse Expression Set 430 | 1429248_PM_at | - | - |
| Mouse Expression Set 430 | 1429248_at | - | - |
| Murine Genome U74v2 Set | 167253_PM_at | - | - |
| Murine Genome U74v2 Set | 167253_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429248_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454309_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454309_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429248_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 794902 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2850092 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850092 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217827 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217827 | - | - |
| mouse-6 v1.1 (Illumina) | scl068020.5_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850092 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068020.5_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850092 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1217827 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217827 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068020.5_23-S | - | - |

Apopt1
Genomic Map