Assay Details
| Gene Symbol: Bphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026512.1 | NP_080788.1 | 5 - 6 | 686 | 132 | |
| GenBank mRNA | AK007606.1 | - | 5 - 6 | 681 | 132 | |
| AK008202.1 | - | 5 - 6 | 705 | 132 | ||
| AK078232.1 | - | 5 - 6 | 731 | 132 | ||
| AK152712.1 | - | 5 - 6 | 696 | 132 | ||
| AK170099.1 | - | 5 - 6 | 689 | 132 | ||
| BC023146.1 | - | 5 - 6 | 671 | 3990118 | 132 | |
| BC025162.1 | - | 5 - 6 | 686 | 4921451 | 132 | |
| BC029322.1 | - | 3 - 4 | 321 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bphl
Genomic Map