Assay Details
| Gene Symbol: Fam92a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026558.4 | NP_080834.3 | 3 - 4 | 615 | 73 | |
| GenBank mRNA | AK013129.1 | - | 3 - 4 | 456 | 73 | |
| AK019574.1 | - | 3 - 4 | 443 | 73 | ||
| AK078438.1 | - | 3 - 4 | 596 | 73 | ||
| AK088705.1 | - | 3 - 4 | 596 | 73 | ||
| BC020162.1 | - | 409 | 3991990 | 73 | ||
| BC055848.1 | - | 3 - 4 | 1451 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam92a
Genomic Map