Assay Details
| Gene Symbol: Agpat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026644.2 | NP_080920.2 | 4 - 5 | 718 | 88 | |
| GenBank mRNA | AK005139.1 | - | 4 - 5 | 719 | 88 | |
| AK166917.1 | - | 4 - 5 | 672 | 88 | ||
| BC031179.1 | - | 4 - 5 | 841 | 88 | ||
| BC047281.1 | - | 4 - 5 | 649 | 6477257 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agpat4
Genomic Map