Assay Details
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177576.1 | NP_001171047.1 | 2 - 3 | 242 | 65 | |
| NM_026646.3 | NP_080922.1 | 2 - 3 | 484 | 65 | ||
| GenBank mRNA | AK010193.1 | - | 2 - 3 | 230 | 65 | |
| AK018760.1 | - | 2 - 3 | 248 | 65 | ||
| AK046874.1 | - | 2 - 3 | 229 | 65 | ||
| AK048389.1 | - | 2 - 3 | 213 | 65 | ||
| AK049313.1 | - | 2 - 3 | 213 | 65 | ||
| AK155276.1 | - | 2 - 3 | 236 | 65 | ||
| AK162400.1 | - | 2 - 3 | 195 | 65 | ||
| BC037949.1 | - | 2 - 3 | 241 | 5367108 | 65 | |
| BC050887.1 | - | 2 - 3 | 437 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK010193.1,AK046874.1,AK048389.1,AK049313.1,AK155276.1,AK162400.1,BC037949.1,BC050887.1,NM_001177576.1,NM_026646.3 transcripts.
Set Membership: |
|

Slc25a22
Genomic Map