Assay Details
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177576.1 | NP_001171047.1 | 5 - 6 | 514 | 67 | |
| NM_026646.3 | NP_080922.1 | 5 - 6 | 756 | 67 | ||
| GenBank mRNA | AK010193.1 | - | 5 - 6 | 502 | 67 | |
| AK046874.1 | - | 5 - 6 | 501 | 67 | ||
| AK048389.1 | - | 5 - 6 | 485 | 67 | ||
| AK049313.1 | - | 5 - 6 | 485 | 67 | ||
| AK155276.1 | - | 5 - 6 | 508 | 67 | ||
| AK162400.1 | - | 5 - 6 | 467 | 67 | ||
| BC037949.1 | - | 5 - 6 | 513 | 5367108 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a22
Genomic Map