Assay Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026647.3 | NP_080923.2 | 5 - 6 | 726 | 66 | |
| GenBank mRNA | AK020293.1 | - | 5 - 6 | 608 | 66 | |
| AY166674.1 | - | 3 - 4 | 364 | 66 | ||
| BC043110.1 | - | 5 - 6 | 572 | 6406930 | 66 | |
| BC058372.1 | - | 5 - 6 | 594 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc21
Genomic Map