Assay Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133925.2 | NP_598686.2 | 15 - 16 | 1804 | 97 | |
| GenBank mRNA | AK156773.1 | - | 15 - 16 | 1741 | 97 | |
| AK161670.1 | - | 15 - 16 | 1771 | 97 | ||
| AK167050.1 | - | 15 - 16 | 1806 | 97 | ||
| BC018373.1 | - | 11 - 12 | 1178 | 3708613 | 97 | |
| BC023293.1 | - | 5 - 6 | 553 | 97 | ||
| BC024602.1 | - | 15 - 16 | 1685 | 97 | ||
| BC027123.1 | - | 8 - 9 | 1110 | 5356457 | 97 | |
| BC040811.1 | - | 15 - 16 | 1749 | 4015830 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm28
Genomic Map