Assay Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133925.2 | NP_598686.2 | 17 - 18 | 2130 | 66 | |
| GenBank mRNA | AK156773.1 | - | 17 - 18 | 2067 | 66 | |
| AK161670.1 | - | 17 - 18 | 2099 | 66 | ||
| AK167050.1 | - | 17 - 18 | 2132 | 66 | ||
| BC018373.1 | - | 13 - 14 | 1504 | 3708613 | 66 | |
| BC023293.1 | - | 7 - 8 | 879 | 66 | ||
| BC024602.1 | - | 17 - 18 | 2011 | 66 | ||
| BC027123.1 | - | 10 - 11 | 1436 | 5356457 | 66 | |
| BC040811.1 | - | 17 - 18 | 2075 | 4015830 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm28
Genomic Map