Assay Details
| Gene Symbol: Pomgnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290658.1 | NP_001277587.1 | 5 - 6 | 600 | 91 | |
| NM_026651.2 | NP_080927.1 | 6 - 7 | 686 | 91 | ||
| NM_029786.2 | NP_084062.2 | 6 - 7 | 686 | 91 | ||
| GenBank mRNA | AB053220.1 | - | 6 - 7 | 668 | 91 | |
| AK002638.1 | - | 6 - 7 | 678 | 91 | ||
| AK015478.1 | - | 5 - 6 | 602 | 91 | ||
| AK019640.1 | - | 5 - 6 | 592 | 91 | ||
| BC011201.1 | - | 6 - 7 | 609 | 4190368 | 91 | |
| BC023265.1 | - | 3 - 4 | 273 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pomgnt1
Genomic Map