Assay Details
| Gene Symbol: Pomgnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290658.1 | NP_001277587.1 | 19 - 20 | 1860 | 59 | |
| NM_026651.2 | NP_080927.1 | 20 - 21 | 1946 | 59 | ||
| NM_029786.2 | NP_084062.2 | 19 - 20 | 1847 | 59 | ||
| GenBank mRNA | AB053220.1 | - | 20 - 21 | 1928 | 59 | |
| AK002638.1 | - | 19 - 20 | 1839 | 59 | ||
| AK015478.1 | - | 19 - 20 | 1862 | 59 | ||
| AK019640.1 | - | 19 - 20 | 1852 | 59 | ||
| AK210391.1 | - | 4 - 5 | 310 | 59 | ||
| AK212205.1 | - | 4 - 5 | 310 | 59 | ||
| BC011201.1 | - | 20 - 21 | 1869 | 4190368 | 59 | |
| BC023265.1 | - | 17 - 18 | 1534 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pomgnt1
Genomic Map