Assay Details
| Gene Symbol: Mcoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005846.2 | NP_001005846.1 | 5 - 6 | 720 | 86 | |
| NM_026656.4 | NP_080932.2 | 5 - 6 | 748 | 86 | ||
| GenBank mRNA | AF503575.1 | - | 5 - 6 | 737 | 86 | |
| AK014467.1 | - | 5 - 6 | 651 | 86 | ||
| AK019454.1 | - | 5 - 6 | 617 | 86 | ||
| AK150446.1 | - | 5 - 6 | 716 | 86 | ||
| AK150547.1 | - | 5 - 6 | 716 | 86 | ||
| AK152067.1 | - | 5 - 6 | 716 | 86 | ||
| AK152943.1 | - | 5 - 6 | 716 | 86 | ||
| AK153290.1 | - | 5 - 6 | 716 | 86 | ||
| AK159172.1 | - | 5 - 6 | 748 | 86 | ||
| AY083532.1 | - | 4 - 5 | 566 | 86 | ||
| BC029847.1 | - | 5 - 6 | 597 | 3967456 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcoln2
Genomic Map