Assay Details
| Gene Symbol: Gsdmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026960.4 | NP_081236.1 | 6 - 7 | 887 | 82 | |
| GenBank mRNA | AK007710.1 | - | 6 - 7 | 888 | 82 | |
| AK088747.1 | - | 6 - 7 | 899 | 82 | ||
| AK165858.1 | - | 6 - 7 | 888 | 82 | ||
| AK171294.1 | - | 6 - 7 | 889 | 82 | ||
| BC029813.1 | - | 6 - 7 | 840 | 4194837 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsdmd
Genomic Map