Assay Details
| Gene Symbol: Rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026967.4 | NP_081243.1 | 3 - 4 | 403 | 73 | |
| GenBank mRNA | AK007708.1 | - | 3 - 4 | 253 | 73 | |
| AK135858.1 | - | 3 - 4 | 358 | 73 | ||
| AY327413.1 | - | 3 - 4 | 451 | 73 | ||
| BC016521.1 | - | 3 - 4 | 322 | 3487009 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhebl1
Genomic Map