Assay Details
| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190357.1 | NP_001177286.1 | 2 - 3 | 279 | 81 | |
| NM_001293661.1 | NP_001280590.1 | 2 - 3 | 279 | 81 | ||
| NM_026995.4 | NP_081271.2 | 2 - 3 | 331 | 81 | ||
| GenBank mRNA | AK003048.1 | - | 2 - 3 | 194 | 81 | |
| AK009254.1 | - | 2 - 3 | 212 | 81 | ||
| AK012456.1 | - | 2 - 3 | 219 | 81 | ||
| AK013028.1 | - | 2 - 3 | 300 | 81 | ||
| AK013069.1 | - | 2 - 3 | 198 | 81 | ||
| AK013420.1 | - | 2 - 3 | 203 | 81 | ||
| AK154142.1 | - | 2 - 3 | 313 | 81 | ||
| AK171034.1 | - | 2 - 3 | 313 | 81 | ||
| BC019538.1 | - | 2 - 3 | 251 | 4211749 | 81 | |
| BC021955.1 | - | 2 - 3 | 192 | 5148177 | 81 | |
| BY270677.1 | - | 2 - 3 | 279 | 81 | ||
| DV045296.1 | - | 2 - 3 | 217 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carkd
Genomic Map