Assay Details
| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190357.1 | NP_001177286.1 | 6 - 7 | 567 | 67 | |
| NM_001293661.1 | NP_001280590.1 | 6 - 7 | 567 | 67 | ||
| NM_026995.4 | NP_081271.2 | 6 - 7 | 619 | 67 | ||
| GenBank mRNA | AK003048.1 | - | 6 - 7 | 482 | 67 | |
| AK009254.1 | - | 6 - 7 | 500 | 67 | ||
| AK012456.1 | - | 6 - 7 | 507 | 67 | ||
| AK013028.1 | - | 6 - 7 | 588 | 67 | ||
| AK013069.1 | - | 6 - 7 | 486 | 67 | ||
| AK013420.1 | - | 6 - 7 | 491 | 67 | ||
| AK154142.1 | - | 6 - 7 | 601 | 67 | ||
| AK171034.1 | - | 6 - 7 | 601 | 67 | ||
| BC019538.1 | - | 6 - 7 | 539 | 4211749 | 67 | |
| BC021955.1 | - | 6 - 7 | 480 | 5148177 | 67 | |
| DV045296.1 | - | 6 - 7 | 505 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carkd
Genomic Map