Assay Details
| Gene Symbol: Lrrc46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027026.2 | NP_081302.2 | 5 - 6 | 663 | 70 | |
| GenBank mRNA | AK005668.1 | - | 5 - 6 | 639 | 70 | |
| AK158517.1 | - | 5 - 6 | 710 | 70 | ||
| BC022723.1 | - | 5 - 6 | 663 | 4503387 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc46
Genomic Map