Assay Details
| Gene Symbol: Lrrc46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027026.2 | NP_081302.2 | 6 - 7 | 725 | 59 | |
| GenBank mRNA | AK005668.1 | - | 6 - 7 | 701 | 59 | |
| AK158517.1 | - | 6 - 7 | 772 | 59 | ||
| BC022723.1 | - | 6 - 7 | 725 | 4503387 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc46
Genomic Map