Assay Details
| Gene Symbol: Lrrc46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027026.2 | NP_081302.2 | 7 - 8 | 874 | 59 | |
| GenBank mRNA | AK005668.1 | - | 7 - 8 | 850 | 59 | |
| AK158517.1 | - | 7 - 8 | 921 | 59 | ||
| BC022723.1 | - | 7 - 8 | 874 | 4503387 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048327 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048328 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | AK158517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | BC022723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | NM_027026.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | AK005668.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | AK158517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | BC022723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | NM_027026.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P375546 | AK005668.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123522 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166408 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | AK158517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | BC022723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | NM_027026.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | AK005668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | AK158517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | BC022723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | NM_027026.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P375546 | AK005668.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048327 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166408 | - | - |
| Whole Mouse Genome G4122A | A_51_P216085 | AK158517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216085 | BC022723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216085 | NM_027026.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216085 | AK005668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | AK158517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | BC022723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | NM_027026.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | AK005668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | AK158517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | BC022723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | NM_027026.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P375546 | AK005668.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117182 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135962 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166408 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09420561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09202623 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09420564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048327 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28048328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09202623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09653034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09491379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09344523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09344519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09653028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09653017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048280 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390325 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424909_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390319 | - | - |
| Mouse Genome 430 2.0 Array | 1424909_at | - | - |
| Mouse Genome 430 2.0 Array | 1424909_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69297_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69297_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100033328.mm.1 | NM_027026.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033328.mm.1 | NM_027026.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100033328.mm.1 | TC1100003634.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424909_at | - | - |
| Mouse Expression Set 430 | 1424909_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424909_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424909_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424909_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424909_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 452218 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4850433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737412 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737412 | - | - |
| mouse-6 v1.1 (Illumina) | scl39656.9.1_122-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39656.9.1_122-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737412 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737412 | - | - |

Lrrc46
Genomic Map