| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00015564.mm.1 |
NM_027109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015564.mm.1 |
NM_027109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015564.mm.1 |
TC0X00002448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015563.mm.1 |
NM_001172154.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015563.mm.1 |
NM_001172154.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015563.mm.1 |
NM_027109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015563.mm.1 |
NM_027109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015563.mm.1 |
TC0X00002448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015565.mm.1 |
NM_027109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015565.mm.1 |
NM_027109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015565.mm.1 |
TC0X00002448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015567.mm.1 |
NM_027109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015567.mm.1 |
NM_027109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015567.mm.1 |
TC0X00002448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015566.mm.1 |
NM_027109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015566.mm.1 |
NM_027109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015566.mm.1 |
TC0X00002448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015562.mm.1 |
NM_001172154.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015562.mm.1 |
NM_001172154.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015562.mm.1 |
NM_027109.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015562.mm.1 |
NM_027109.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015562.mm.1 |
TC0X00002448.mm.1 |
MTA TC |