Assay Details
| Gene Symbol: Dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172154.1 | NP_001165625.1 | 5 - 6 | 666 | 60 | |
| NM_027109.2 | NP_081385.1 | 5 - 6 | 783 | 60 | ||
| GenBank mRNA | AK009174.1 | - | 5 - 6 | 789 | 60 | |
| AK150242.1 | - | 5 - 6 | 696 | 60 | ||
| BC023246.1 | - | 5 - 6 | 666 | 4039470 | 60 | |
| DQ116784.1 | - | 5 - 6 | 677 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1l1
Genomic Map