Assay Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027122.3 | NP_081398.2 | 2 - 3 | 957 | 81 | |
| GenBank mRNA | AK009269.1 | - | 2 - 3 | 1036 | 81 | |
| AK089539.1 | - | 2 - 3 | 958 | 81 | ||
| AK217168.1 | - | 2 - 3 | 385 | 81 | ||
| BC019171.1 | - | 2 - 3 | 984 | 5003354 | 81 | |
| BC085305.1 | - | 2 - 3 | 1021 | 6399056 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd3
Genomic Map