| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900006437.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006437.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006437.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006434.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006434.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006434.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006439.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006439.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006439.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006442.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006442.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006442.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006438.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006438.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006438.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006441.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006441.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006441.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006432.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006432.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006432.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006440.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006440.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006440.mm.1 |
TC0900000844.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006444.mm.1 |
NM_027134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006444.mm.1 |
NM_027134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006444.mm.1 |
TC0900000844.mm.1 |
MTA TC |