Assay Details
| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010419.1 | - | 2 - 3 | 335 | 116 | |
| BC022648.1 | - | 2 - 3 | 296 | 116 | ||
| BC117053.1 | - | 2 - 3 | 321 | 116 | ||
| BC117055.1 | - | 2 - 3 | 321 | 116 | ||
More Information
Set Membership: |
|

Lrr1
Genomic Map