Assay Details
| Gene Symbol: Trmt10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027266.4 | NP_081542.2 | 6 - 7 | 714 | 73 | |
| GenBank mRNA | AK011746.1 | - | 6 - 7 | 727 | 73 | |
| AK030465.1 | - | 6 - 7 | 715 | 73 | ||
| BC019428.1 | - | 5 - 6 | 691 | 4205772 | 73 | |
More Information
Set Membership: |
|

Trmt10b
Genomic Map