Assay Details
Gene Symbol: Rpp25l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027278.3 | NP_081554.2 | 1 - 2 | 57 | 59 | |
GenBank mRNA | AJ293897.1 | - | 1 - 2 | 52 | 59 | |
AK013222.1 | - | 1 - 2 | 46 | 59 | ||
BC047068.1 | - | 1 - 2 | 49 | 5009034 | 59 | |
BF146902.1 | - | 1 - 2 | 57 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AJ293897.1,AK013222.1,BC047068.1,NM_027278.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863909 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863910 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863906 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863908 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | NM_027278.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | AJ293897.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | AK013222.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | BC047068.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | NM_027278.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | AJ293897.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | AK013222.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411130 | BC047068.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522647 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173357 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | NM_027278.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | AJ293897.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | AK013222.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | BC047068.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | NM_027278.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | AJ293897.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | AK013222.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411130 | BC047068.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863901 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863895 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863912 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863897 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22863907 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173357 | - | - |
Whole Mouse Genome G4122A | A_51_P411130 | NM_027278.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | AJ293897.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | AK013222.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | BC047068.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | NM_027278.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | AJ293897.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | AK013222.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P411130 | BC047068.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P429106 | NM_027278.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P429106 | AJ293897.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P429106 | AK013222.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P429106 | BC047068.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118826 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173357 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03439026 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03439013 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03439018 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03370875 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03439021 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863910 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863909 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863906 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22863908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22863872 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863913 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03761408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03370873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03370865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03808089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03808107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03808084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03761412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439036 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03483605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03808099 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439021 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22863866 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10512289 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512290 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427085_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10512288 | - | - |
Mouse Genome 430 2.0 Array | 1427085_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427085_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69961_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69961_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400020001.mm.1 | NM_027278.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400020001.mm.1 | NM_027278.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400020001.mm.1 | TC0400002476.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400020002.mm.1 | NM_027278.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400020002.mm.1 | NM_027278.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400020002.mm.1 | TC0400002476.mm.1 | MTA TC |
Murine Genome U74v2 Set | 102194_at | - | - |
Murine Genome U74v2 Set | 102194_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427085_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427085_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 813192 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2900463 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2900463 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002900463 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732677 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2732677 | - | - |
mouse-6 v1.1 (Illumina) | scl24416.3.1_12-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2900463 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl24416.3.1_12-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900463 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2974601 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2974601 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24416.3.1_12-S | - | - |
