Assay Details
| Gene Symbol: Rpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027371.3 | NP_081647.2 | 6 - 7 | 726 | 102 | |
| GenBank mRNA | AK010079.1 | - | 6 - 7 | 723 | 102 | |
| AK162073.1 | - | 6 - 7 | 724 | 102 | ||
| AK171079.1 | - | 6 - 7 | 715 | 102 | ||
| BC056165.1 | - | 6 - 7 | 677 | 5694049 | 102 | |
| BC089369.1 | - | 6 - 7 | 711 | 30550759 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpf1
Genomic Map