Assay Details
| Gene Symbol: Rpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027371.3 | NP_081647.2 | 8 - 9 | 1040 | 122 | |
| GenBank mRNA | AK010079.1 | - | 8 - 9 | 1037 | 122 | |
| AK162073.1 | - | 8 - 9 | 1039 | 122 | ||
| AK171079.1 | - | 8 - 9 | 1029 | 122 | ||
| BC056165.1 | - | 8 - 9 | 991 | 5694049 | 122 | |
| BC089369.1 | - | 8 - 9 | 1025 | 30550759 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpf1
Genomic Map