Assay Details
| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027439.4 | NP_081715.1 | 2 - 3 | 312 | 75 | |
| GenBank mRNA | AB192471.2 | - | 2 - 3 | 188 | 75 | |
| AK017482.1 | - | 2 - 3 | 244 | 75 | ||
| AK029405.1 | - | 2 - 3 | 215 | 75 | ||
| AK076495.1 | - | 2 - 3 | 257 | 75 | ||
| AK151735.1 | - | 2 - 3 | 243 | 75 | ||
| AK168139.1 | - | 2 - 3 | 271 | 75 | ||
| BC014706.1 | - | 2 - 3 | 183 | 4457079 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6ap2
Genomic Map