Assay Details
| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027439.4 | NP_081715.1 | 6 - 7 | 729 | 69 | |
| GenBank mRNA | AB192471.2 | - | 6 - 7 | 605 | 69 | |
| AK017482.1 | - | 6 - 7 | 661 | 69 | ||
| AK029405.1 | - | 6 - 7 | 632 | 69 | ||
| AK076495.1 | - | 6 - 7 | 674 | 69 | ||
| AK151735.1 | - | 6 - 7 | 661 | 69 | ||
| AK168139.1 | - | 6 - 7 | 686 | 69 | ||
| AW985491.1 | - | 3 - 4 | 296 | 69 | ||
| BC014706.1 | - | 6 - 7 | 600 | 4457079 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6ap2
Genomic Map