Assay Details
| Gene Symbol: Tmem144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027495.4 | NP_081771.2 | 2 - 3 | 440 | 69 | |
| GenBank mRNA | AK017808.1 | - | 2 - 3 | 459 | 69 | |
| AK079384.1 | - | 2 - 3 | 441 | 69 | ||
| BC018493.1 | - | 2 - 3 | 348 | 3593194 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem144
Genomic Map