Assay Details
| Gene Symbol: Tmem144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027495.4 | NP_081771.2 | 7 - 8 | 907 | 96 | |
| GenBank mRNA | AK017808.1 | - | 7 - 8 | 926 | 96 | |
| AK079384.1 | - | 7 - 8 | 908 | 96 | ||
| BC018493.1 | - | 7 - 8 | 815 | 3593194 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem144
Genomic Map